8-Fluoroquinoline-5-carboxylic acid 96%

Supplier: Apollo Scientific

PC904283-1G PC904283-250MG
APOSPC904283-1GEA 877 EUR
APOSPC904283-1G APOSPC904283-250MG
8-Fluoroquinoline-5-carboxylic acid 96%
8-Fluoroquinoline-5-carboxylic acid
Formula: C₁₀H₆FNO₂
MW: 191,159 g/mol
Storage Temperature: Ambient
MDL Number: MFCD20658731
CAS Number: 204782-93-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR