2-Chloro-6-fluoroquinazolin-4(3H)-one 95%

Supplier: Apollo Scientific

PC904676-1G PC904676-250MG
APOSPC904676-1GEA 683 EUR
APOSPC904676-1G APOSPC904676-250MG
2-Chloro-6-fluoroquinazolin-4(3H)-one 95%
2-Chloro-6-fluoroquinazolin-4(3H)-one
Formula: C₈H₄ClFN₂O
MW: 198,582 g/mol
Storage Temperature: Ambient
MDL Number: MFCD11520685
CAS Number: 769158-12-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR