5-(Chloromethyl)-2-fluoropyridine 96%

Supplier: Apollo Scientific

PC904728-1G PC904728-250MG
APOSPC904728-1GEA 452 EUR
APOSPC904728-1G APOSPC904728-250MG
5-(Chloromethyl)-2-fluoropyridine 96%
5-(Chloromethyl)-2-fluoropyridine
Formula: C₆H₅ClFN
MW: 145,562 g/mol
Storage Temperature: Ambient
MDL Number: MFCD10697563
CAS Number: 315180-15-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR