4-Amino-1,1,1-trifluoro-pent-3-en-2-one 95%
Supplier: Apollo Scientific
PC904754-1G
PC904754-250MG
APOSPC904754-1GEA
464
EUR
APOSPC904754-1G
APOSPC904754-250MG
4-Amino-1,1,1-trifluoro-pent-3-en-2-one 95%
4-Amino-1,1,1-trifluoro-pent-3-en-2-one
Formula:
C₅H₆F₃NO Storage Temperature: Ambient |
MDL Number:
MFCD00230875 CAS Number: 72885-02-0 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...