2-Amino-4-chloro-6-(trifluoromethyl)pyrimidine 98%
Supplier: Apollo Scientific
PC904758-10G
PC904758-1G
PC904758-5G
APOSPC904758-10GEA
357
EUR
APOSPC904758-10G
APOSPC904758-1G
APOSPC904758-5G
2-Amino-4-chloro-6-(trifluoromethyl)pyrimidine 98%
2-Amino-4-chloro-6-(trifluoromethyl)pyrimidine
Formula:
C₅H₃ClF₃N₃ MW: 197,546 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD06656426 CAS Number: 16097-60-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...