5-Fluoro-3,4-dihydronaphthalen-1(2H)-one 97%
Supplier: Apollo Scientific
|
Warning
|
PC904824-100MG
PC904824-1G
PC904824-250MG
APOSPC904824-100MGEA
94.4
EUR
APOSPC904824-100MG
APOSPC904824-1G
APOSPC904824-250MG
5-Fluoro-3,4-dihydronaphthalen-1(2H)-one 97%
5-Fluoro-3,4-dihydronaphthalen-1(2H)-one
Formula:
C₁₀H₉FO MW: 164,176 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD08234371 CAS Number: 93742-85-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...