5,6-Difluoro-1H-indazole-3-carboxylic acid 95%

Supplier: Apollo Scientific

PC905086-100MG PC905086-1G PC905086-250MG
APOSPC905086-100MGEA 132 EUR
APOSPC905086-100MG APOSPC905086-1G APOSPC905086-250MG
5,6-Difluoro-1H-indazole-3-carboxylic acid 95%
5,6-Difluoro-1H-indazole-3-carboxylic acid
Formula: C₈H₄F₂N₂O₂
Storage Temperature: Ambient
MDL Number: MFCD05663995
CAS Number: 129295-33-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR