5-Chloro-3-(chloromethyl)-2-fluoropyridine 95%

Supplier: Apollo Scientific

PC905200-1G PC905200-250MG
APOSPC905200-1GEA 633 EUR
APOSPC905200-1G APOSPC905200-250MG
5-Chloro-3-(chloromethyl)-2-fluoropyridine 95%
5-Chloro-3-(chloromethyl)-2-fluoropyridine
Formula: C₆H₄Cl₂FN
MW: 180,007 g/mol
Storage Temperature: Ambient
MDL Number: MFCD16608118
CAS Number: 1227606-17-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR