2-(2-Fluorophenyl)-4,4-dimethyl-4,5-dihydro-1,3-oxazole 95%

Supplier: Apollo Scientific

PC905223-1G PC905223-250MG
APOSPC905223-1GEA 864 EUR
APOSPC905223-1G APOSPC905223-250MG
2-(2-Fluorophenyl)-4,4-dimethyl-4,5-dihydro-1,3-oxazole 95%
2-(2-Fluorophenyl)-4,4-dimethyl-4,5-dihydro-1,3-oxazole
Formula: C₁₁H₁₂FNO
MW: 193,217 g/mol
Storage Temperature: Ambient
MDL Number: MFCD04108264
CAS Number: 66464-20-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR