3-(3-Fluorophenyl)tetrahydro-3-furancarbaldehyde 95%

Supplier: Apollo Scientific

PC905224-1G PC905224-250MG
APOSPC905224-1GEA 712 EUR
APOSPC905224-1G APOSPC905224-250MG
3-(3-Fluorophenyl)tetrahydro-3-furancarbaldehyde 95%
3-(3-Fluorophenyl)tetrahydro-3-furancarbaldehyde
Formula: C₁₁H₁₁FO₂
Storage Temperature: Ambient
MDL Number: MFCD11053959
CAS Number: 1060817-40-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR