2-Fluoro-6-methylbenzamide 98%

Supplier: Apollo Scientific

PC905296-1G PC905296-250MG PC905296-5G
APOSPC905296-1GEA 255 EUR
APOSPC905296-1G APOSPC905296-250MG APOSPC905296-5G
2-Fluoro-6-methylbenzamide 98%
2-Fluoro-6-methylbenzamide
Formula: C₈H₈FNO
MW: 153,154 g/mol
Storage Temperature: Ambient
MDL Number: MFCD06660279
CAS Number: 886502-14-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR