Potassium (chloromethyl)trifluoroborate 95%

Supplier: Apollo Scientific
Warning

PC905379-1G PC905379-5G
APOSPC905379-1GEA 48 EUR
APOSPC905379-1G APOSPC905379-5G
Potassium (chloromethyl)trifluoroborate 95%
Potassium (chloromethyl)trifluoroborate
Formula: CH₂BClF₃·K
MW: 156,384 g/mol
Storage Temperature: Freezer
MDL Number: MFCD11052821
CAS Number: 1279123-64-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR