3-Fluoro-5-nitropyridine 95%

Supplier: Apollo Scientific

PC905559-1G PC905559-250MG PC905559-5G
APOSPC905559-1GEA 154 EUR
APOSPC905559-1G APOSPC905559-250MG APOSPC905559-5G
3-Fluoro-5-nitropyridine 95%
3-Fluoro-5-nitropyridine
Formula: C₅H₃FN₂O₂
MW: 142,088 g/mol
Storage Temperature: Ambient
MDL Number: MFCD13189114
CAS Number: 1060804-39-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR