5-Bromo-4-fluoroindoline 95%

Supplier: Apollo Scientific
Warning

PC905636-100MG PC905636-250MG
APOSPC905636-100MGEA 66.7 EUR
APOSPC905636-100MG APOSPC905636-250MG
5-Bromo-4-fluoroindoline 95%
5-Bromo-4-fluoroindoline
Formula: C₈H₇BrFN
MW: 216,05 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD27991352
CAS Number: 1782463-27-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR