6-(Trifluoromethyl)-1-indanone 98%
Supplier: Apollo Scientific
PC906472-1G
PC906472-250MG
APOSPC906472-1GEA
309
EUR
APOSPC906472-1G
APOSPC906472-250MG
6-(Trifluoromethyl)-1-indanone 98%
6-(Trifluoromethyl)-1-indanone
Formula:
C₁₀H₇F₃O MW: 200,157 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD07772123 CAS Number: 68755-37-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...