2,6-Difluorobenzene-1-carbothioamide 95%

Supplier: Apollo Scientific

PC906491-1G PC906491-250MG
APOSPC906491-1GEA 488 EUR
APOSPC906491-1G APOSPC906491-250MG
2,6-Difluorobenzene-1-carbothioamide 95%
2,6-Difluorobenzene-1-carbothioamide
Formula: C₇H₅F₂NS
MW: 173,183 g/mol
Storage Temperature: Ambient
MDL Number: MFCD09809511
CAS Number: 60230-33-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR