7-Fluoro-3,4-dihydroquinolin-2(1H)-one ≥98%

Supplier: Apollo Scientific

PC906505-1G PC906505-250MG
APOSPC906505-1GEA 267 EUR
APOSPC906505-1G APOSPC906505-250MG
7-Fluoro-3,4-dihydroquinolin-2(1H)-one ≥98%
7-Fluoro-3,4-dihydroquinolin-2(1H)-one
Formula: C₉H₈FNO
MW: 165,164 g/mol
Storage Temperature: Ambient
MDL Number: MFCD11811768
CAS Number: 4590-52-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR