3-Bromo-4-fluoro-5-trifluoromethyl-phenylamine 97%
Supplier: Apollo Scientific
PC906734-1G
PC906734-5G
PC906734-25G
APOSPC906734-1GEA
27.7
EUR
APOSPC906734-1G
APOSPC906734-5G
APOSPC906734-25G
3-Bromo-4-fluoro-5-trifluoromethyl-phenylamine 97%
3-Bromo-4-fluoro-5-trifluoromethyl-phenylamine
Formula:
C₇H₄BrF₄N Storage Temperature: Ambient |
MDL Number:
MFCD11520114 CAS Number: 1233026-11-3 UN: 1759 ADR: 8,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...