1-(2-Amino-4-fluoro-5-methoxyphenyl)ethanone 95%

Supplier: Apollo Scientific

PC906992-5G PC906992-1G
APOSPC906992-5GEA 0 EUR
APOSPC906992-5G APOSPC906992-1G
1-(2-Amino-4-fluoro-5-methoxyphenyl)ethanone 95%
1-(2-Amino-4-fluoro-5-methoxyphenyl)ethanone
Formula: C₉H₁₀FNO₂
MW: 183,18 g/mol
Storage Temperature: Ambient
MDL Number: MFCD11113349
CAS Number: 949159-97-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR