N-(2,5-Difluoro-phenyl)-formamide 95%
Supplier: Apollo Scientific
PC907184-1G
PC907184-250MG
APOSPC907184-1GEA
574
EUR
APOSPC907184-1G
APOSPC907184-250MG
N-(2,5-Difluoro-phenyl)-formamide 95%
N-(2,5-Difluoro-phenyl)-formamide
Formula:
C₇H₅F₂NO Storage Temperature: Ambient |
MDL Number:
MFCD21121183 CAS Number: 81183-56-4 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...