Methyl 3-amino-5-fluoro-4-methylbenzoate 95%

Supplier: Apollo Scientific

PC907212-1G PC907212-250MG
APOSPC907212-1GEA 793 EUR
APOSPC907212-1G APOSPC907212-250MG
Methyl 3-amino-5-fluoro-4-methylbenzoate 95%
Methyl 3-amino-5-fluoro-4-methylbenzoate
Formula: C₉H₁₀FNO₂
MW: 183,18 g/mol
Storage Temperature: Ambient
MDL Number: MFCD14694719
CAS Number: 1093087-06-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR