2-(2,5-Difluorophenoxy)acetic acid 95%
Supplier: Apollo Scientific
PC907340-1G
PC907340-250MG
APOSPC907340-1GEA
539
EUR
APOSPC907340-1G
APOSPC907340-250MG
2-(2,5-Difluorophenoxy)acetic acid 95%
2-(2,5-Difluorophenoxy)acetic acid
Formula:
C₈H₆F₂O₃ MW: 188,128 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD06823826 CAS Number: 366-56-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...