5-Fluoro-2-methoxyisonicotinaldehyde 95%
Supplier: Apollo Scientific
PC907873-1G
PC907873-250MG
APOSPC907873-1GEA
575
EUR
APOSPC907873-1G
APOSPC907873-250MG
5-Fluoro-2-methoxyisonicotinaldehyde 95%
5-Fluoro-2-methoxyisonicotinaldehyde
Formula:
C₇H₆FNO₂ MW: 155,126 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD08277260 CAS Number: 884495-12-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...