7-Fluoroisoquinoline ≥98%

Supplier: Apollo Scientific

PC907907-100MG PC907907-1G
APOSPC907907-100MGEA 85.7 EUR
APOSPC907907-100MG APOSPC907907-1G
7-Fluoroisoquinoline ≥98%
7-Fluoroisoquinoline
Formula: C₉H₆FN
Storage Temperature: Ambient
MDL Number: MFCD11226905
CAS Number: 1075-12-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR