6-Amino-2,3-difluorobenzonitrile 95%

Supplier: Apollo Scientific

PC908023-1G PC908023-5G
APOSPC908023-1GEA 292 EUR
APOSPC908023-1G APOSPC908023-5G
6-Amino-2,3-difluorobenzonitrile 95%
6-Amino-2,3-difluorobenzonitrile
Formula: C₇H₄F₂N₂
MW: 154,117 g/mol
Storage Temperature: Ambient
MDL Number: MFCD22490563
CAS Number: 1805635-10-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR