1-(2,2,2-Trifluoroethyl)azetidin-3-amine
Supplier: Apollo Scientific
PC908306-1G
PC908306-250MG
PC908306-500MG
APOSPC908306-1GEA
1080
EUR
APOSPC908306-1G
APOSPC908306-250MG
APOSPC908306-500MG
1-(2,2,2-Trifluoroethyl)azetidin-3-amine
1-(2,2,2-Trifluoroethyl)azetidin-3-amine
Formula:
C₅H₉F₃N₂ MW: 154,134 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD18882621 CAS Number: 1339236-05-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...