5-Fluorobenzofuran-4-carbaldehyde ≥98%

Supplier: Apollo Scientific

PC908480-100MG PC908480-1G PC908480-250MG
APOSPC908480-100MGEA 111 EUR
APOSPC908480-100MG APOSPC908480-1G APOSPC908480-250MG
5-Fluorobenzofuran-4-carbaldehyde ≥98%
5-Fluorobenzofuran-4-carbaldehyde
Formula: C₉H₅FO₂
MW: 164,133 g/mol
Storage Temperature: Ambient
MDL Number: MFCD18819778
CAS Number: 199391-70-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR