2-Bromo-6-fluoropyridin-3-ol 95%

Supplier: Apollo Scientific

PC908484-1G PC908484-250MG
APOSPC908484-1GEA 33.5 EUR
APOSPC908484-1G APOSPC908484-250MG
2-Bromo-6-fluoropyridin-3-ol 95%
2-Bromo-6-fluoropyridin-3-ol
Formula: C₅H₃BrFNO
MW: 191,986 g/mol
Storage Temperature: Ambient
MDL Number: MFCD18257625
CAS Number: 1256822-94-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR