4-Fluorobenzofuran-7-carbaldehyde 95%
Supplier: Apollo Scientific
PC908731-100MG
PC908731-1G
PC908731-250MG
APOSPC908731-100MGEA
168
EUR
APOSPC908731-100MG
APOSPC908731-1G
APOSPC908731-250MG
4-Fluorobenzofuran-7-carbaldehyde 95%
4-Fluorobenzofuran-7-carbaldehyde
Formula:
C₉H₅FO₂ Storage Temperature: Ambient |
MDL Number:
MFCD11052336 CAS Number: 555155-07-2 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...