5-Fluoro-2-hydroxypyrimidine 95%
Supplier: Apollo Scientific
PC908749-1G
APOSPC908749-1GEA
46.4
EUR
APOSPC908749-1G
5-Fluoro-2-hydroxypyrimidine 95%
5-Fluoro-2-hydroxypyrimidine
Formula:
C₄H₃FN₂O MW: 114,078 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD00223678 CAS Number: 214290-48-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...