5-(trifluoromethyl)-1H-benzo-1,2,3-triazole 95%

Supplier: Apollo Scientific

PC909150-1G PC909150-5G PC909150-10G
APOSPC909150-1GEA 168 EUR
APOSPC909150-1G APOSPC909150-5G APOSPC909150-10G
5-(trifluoromethyl)-1H-benzo-1,2,3-triazole 95%
5-(trifluoromethyl)-1H-benzo-1,2,3-triazole
Formula: C₇H₄F₃N₃
MW: 187,122 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00173974
CAS Number: 1548-67-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR