2-Chloro-4-fluoro-6-nitroaniline 96%
Supplier: Apollo Scientific
PC909152-1G
PC909152-5G
APOSPC909152-1GEA
219
EUR
APOSPC909152-1G
APOSPC909152-5G
2-Chloro-4-fluoro-6-nitroaniline 96%
2-Chloro-4-fluoro-6-nitroaniline
Formula:
C₆H₄ClFN₂O₂ MW: 190,56 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD22571067 CAS Number: 153505-32-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...