3-Fluoro-5-iodobenzaldehyde

Supplier: Apollo Scientific
Warning

PC9096-1G PC9096-5G
APOSPC9096-1GEA 169 EUR
APOSPC9096-1G APOSPC9096-5G
3-Fluoro-5-iodobenzaldehyde
3-Fluoro-5-iodobenzaldehyde
Formula: C₇H₄FIO
MW: 250,009 g/mol
Storage Temperature: Ambient
MDL Number: MFCD07779030
CAS Number: 914636-93-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR