Methyl 2-(2-fluorophenyl)-2-methylpropanoate 95%

Supplier: Apollo Scientific

PC909746-1G PC909746-250MG
APOSPC909746-1GEA 632 EUR
APOSPC909746-1G APOSPC909746-250MG
Methyl 2-(2-fluorophenyl)-2-methylpropanoate 95%
Methyl 2-(2-fluorophenyl)-2-methylpropanoate
Formula: C₁₁H₁₃FO₂
MW: 196,218 g/mol
Storage Temperature: Ambient
MDL Number: MFCD13184250
CAS Number: 861901-19-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR