4-Fluoro-2-(hydroxymethyl)phenol 95%

Supplier: Apollo Scientific

PC910230-1G PC910230-5G
APOSPC910230-1GEA 219 EUR
APOSPC910230-1G APOSPC910230-5G
4-Fluoro-2-(hydroxymethyl)phenol 95%
4-Fluoro-2-(hydroxymethyl)phenol
Formula: C₇H₇FO₂
MW: 142,128 g/mol
Storage Temperature: Ambient
MDL Number: MFCD03412237
CAS Number: 2357-33-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR