5-Bromo-7-fluorochroman-4-one 95+%

Supplier: Apollo Scientific
Warning

PC910388-100MG PC910388-250MG
APOSPC910388-100MGEA 46.4 EUR
APOSPC910388-100MG APOSPC910388-250MG
5-Bromo-7-fluorochroman-4-one 95+%
5-Bromo-7-fluorochroman-4-one
Formula: C₉H₆BrFO₂
MW: 245.045 g/mol
Storage Temperature: Ambient
MDL Number: MFCD18208093
CAS Number: 1260008-29-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR