8-Bromo-2-fluoro-1,5-naphthyridine 97%
Supplier: Apollo Scientific
|
Warning
|
PC910430-100MG
PC910430-250MG
PC910430-1G
APOSPC910430-100MGEA
332
EUR
APOSPC910430-100MG
APOSPC910430-250MG
APOSPC910430-1G
8-Bromo-2-fluoro-1,5-naphthyridine 97%
8-Bromo-2-fluoro-1,5-naphthyridine
Formula:
C₈H₄BrFN₂ MW: 227,033 g/mol Storage Temperature: Refrigerator |
MDL Number:
MFCD24539414 CAS Number: 1432322-85-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...