Methyl 6-fluoro-1H-indazole-3-carboxylate 95%

Supplier: Apollo Scientific

PC910615-1G
APOSPC910615-1GEA 36.4 EUR
APOSPC910615-1G
Methyl 6-fluoro-1H-indazole-3-carboxylate 95%
Methyl 6-fluoro-1H-indazole-3-carboxylate
Formula: C₉H₇FN₂O₂
MW: 194,162 g/mol
Storage Temperature: Ambient
MDL Number: MFCD07371604
CAS Number: 885279-26-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR