2-Fluoro-4-(pyrrolidin-1-yl)pyridine 98%
Supplier: Apollo Scientific
PC910693-1G
PC910693-5G
APOSPC910693-1GEA
219
EUR
APOSPC910693-1G
APOSPC910693-5G
2-Fluoro-4-(pyrrolidin-1-yl)pyridine 98%
2-Fluoro-4-(pyrrolidin-1-yl)pyridine
Formula:
C₉H₁₁FN₂ MW: 166,195 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD20491454 CAS Number: 1352318-60-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...