3-(Chloromethyl)-2-fluoropyridine 98%
Supplier: Apollo Scientific
PC910721-1G
PC910721-5G
APOSPC910721-1GEA
292
EUR
APOSPC910721-1G
APOSPC910721-5G
3-(Chloromethyl)-2-fluoropyridine 98%
3-(Chloromethyl)-2-fluoropyridine
Formula:
C₆H₅ClFN MW: 145,562 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD10697617 CAS Number: 315180-14-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...