6-Amino-3-bromo-2-fluoropyridine 98%

Supplier: Apollo Scientific

Synonyms: 5-Bromo-6-fluoropyridin-2-amine

PC910727-1G PC910727-250MG
APOSPC910727-1GEA 115 EUR
APOSPC910727-1G APOSPC910727-250MG
6-Amino-3-bromo-2-fluoropyridine 98%
6-Amino-3-bromo-2-fluoropyridine
Formula: C₅H₄BrFN₂
MW: 191,001 g/mol
Storage Temperature: Ambient
MDL Number: MFCD11848480
CAS Number: 944401-65-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR