N-[(3-Fluorophenyl)methyl]cyclopentanamine 98%

Supplier: Apollo Scientific

PC910857-1G PC910857-5G
APOSPC910857-1GEA 219 EUR
APOSPC910857-1G APOSPC910857-5G
N-[(3-Fluorophenyl)methyl]cyclopentanamine 98%
N-[(3-Fluorophenyl)methyl]cyclopentanamine
Formula: C₁₂H₁₆FN
MW: 193,261 g/mol
Storage Temperature: Ambient
MDL Number: MFCD08674991
CAS Number: 85952-78-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR