5-(4-Fluorophenyl)-3-methyl-1,2,4-oxadiazole 98%
Supplier: Apollo Scientific
PC910935-1G
PC910935-5G
APOSPC910935-1GEA
232
EUR
APOSPC910935-1G
APOSPC910935-5G
5-(4-Fluorophenyl)-3-methyl-1,2,4-oxadiazole 98%
5-(4-Fluorophenyl)-3-methyl-1,2,4-oxadiazole
Formula:
C₉H₇FN₂O MW: 178,163 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD09261229 CAS Number: 59562-68-4 |
Learn more
![](/m-ie.vwr.com/en_IE/images/about_vwr_275x315.jpg)
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...