5-Fluoro-1-methylindazole 96%

Supplier: Apollo Scientific

PC910940-1G PC910940-250MG
APOSPC910940-1GEA 632 EUR
APOSPC910940-1G APOSPC910940-250MG
5-Fluoro-1-methylindazole 96%
5-Fluoro-1-methylindazole
Formula: C₈H₇FN₂
MW: 150,153 g/mol
Storage Temperature: Ambient
MDL Number: MFCD11977520
CAS Number: 121002-36-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR