3-Bromo-6-fluoropyridin-2-amine 96%
Supplier: Apollo Scientific
|
Warning
|
PC911050-100MG
PC911050-250MG
APOSPC911050-100MGEA
78.6
EUR
APOSPC911050-100MG
APOSPC911050-250MG
3-Bromo-6-fluoropyridin-2-amine 96%
3-Bromo-6-fluoropyridin-2-amine
Formula:
C₅H₄BrFN₂ MW: 191,001 g/mol Storage Temperature: Refrigerator |
MDL Number:
MFCD16556245 CAS Number: 1232431-41-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...