N-ortho-Fluorophenylglycine 98%
Supplier: Apollo Scientific
PC911097-1G
PC911097-5G
APOSPC911097-1GEA
168
EUR
APOSPC911097-1G
APOSPC911097-5G
N-ortho-Fluorophenylglycine 98%
N-ortho-Fluorophenylglycine
Formula:
C₈H₈FNO₂ Storage Temperature: Ambient |
MDL Number:
MFCD00002687 CAS Number: 5319-42-6 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...