2-(Trifluoroacetyl)pyridine 96%

Supplier: Apollo Scientific

PC911318-1G PC911318-5G PC911318-10G
APOSPC911318-1GEA 190 EUR
APOSPC911318-1G APOSPC911318-5G APOSPC911318-10G
2-(Trifluoroacetyl)pyridine 96%
2-(Trifluoroacetyl)pyridine
Formula: C₇H₄F₃NO
MW: 175,108 g/mol
Storage Temperature: Ambient
MDL Number: MFCD07380748
CAS Number: 33284-17-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR