3-Methoxy-2,4,5-trifluorobenzylamine 95%

Supplier: Apollo Scientific

PC911358-1G PC911358-250MG
APOSPC911358-1GEA 864 EUR
APOSPC911358-1G APOSPC911358-250MG
3-Methoxy-2,4,5-trifluorobenzylamine 95%
3-Methoxy-2,4,5-trifluorobenzylamine
Formula: C₈H₈F₃NO
MW: 191,15 g/mol
Storage Temperature: Ambient
MDL Number: MFCD03839930
CAS Number: 886362-79-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR