8-Fluoro-2,4(1h,3H)-quinazolinedione 97%

Supplier: Apollo Scientific

Synonyms: 8-Fluoroquinazoline-2,4(1H,3H)-dione

PC911372-1G PC911372-5G
APOSPC911372-1GEA 256 EUR
APOSPC911372-1G APOSPC911372-5G
8-Fluoro-2,4(1h,3H)-quinazolinedione 97%
8-Fluoro-2,4(1h,3H)-quinazolinedione
Formula: C₈H₅FN₂O₂
MW: 180,136 g/mol
Storage Temperature: Ambient
MDL Number: MFCD09954844
CAS Number: 959236-96-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR