4-(Aminomethyl)-2-fluorobenzonitrile ≥98%
Supplier: Apollo Scientific
PC911412-100MG
PC911412-250MG
APOSPC911412-100MGEA
578
EUR
APOSPC911412-100MG
APOSPC911412-250MG
4-(Aminomethyl)-2-fluorobenzonitrile ≥98%
4-(Aminomethyl)-2-fluorobenzonitrile
Formula:
C₈H₇FN₂ MW: 150,153 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD06658314 CAS Number: 368426-73-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...